Learn More
Macherey-Nagel™ Nucleosil™ 300-10 Anion II HPLC CC Guard Columns
Product Code. 10449161 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Each
Product Code. Diameter (Metric) Inner unitSize
10449161 4 mm Each
1 missing translation for 'options'
Product Code. 10449161
missing translation for 'mfr': Macherey-Nagel™ 721452.40
Only null left
Specifications
pH 2 to 7.5
Particle Size 10 μm
Length (Metric) 8 mm
Pore Size 300Å
Type HPLC CC Guard Column
Diameter (Metric) Inner 4 mm
Stationary Phase NUCLEOSIL™ Anion II
Mobile Phase Compatibility Diammonium Hydrogen Phosphate Buffer
Solvents Diammonium Hydrogen Phosphate Buffer
Packing Material Silica
Column Format Guard Column
For Use With (Application) Detection of Conductivity or Negative UV Detection
Technique HPLC
Particle Shape Spherical