Learn More
Macherey-Nagel™ Nucleosil™ C4 CC Guard Columns
Product Code. 12738142 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
12738142 4 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12738142 Supplier Macherey-Nagel™ Supplier No. 721889.40
Only null left
Specifications
Carbon Load ∽ 4%
pH 2 to 8
Particle Size 5 μm
Length (Metric) 8 mm
Pore Size 120Å
Type HPLC CC Guard Column
Diameter (Metric) Inner 4 mm
Stationary Phase NUCLEOSIL™ C4
Mobile Phase Compatibility Butyl
Solvents Butyl
Packing Material Silica
Crosslinking Butyl
Column Format Guard Column
For Use With (Application) RP and Ion Pairing chromatography
Technique HPLC
Particle Shape Spherical
Hydrophobicity Yes
Functionality/Interaction Type Hydrophobic
Endcapped Yes
USP Type L26