Learn More
Macherey-Nagel™ Nucleosil™ Chiral-1 CC Guard Columns Cartridges
Brand: Macherey-Nagel™ 721455.40
Additional Details : Weight : 0.01000kg
Specifica
1 Pack | |
8 mm | |
4 mm | |
NUCLEOSIL™ Chiral-1 |
5 μm | |
CC Guard Columns Cartridges | |
EC columns of 4 and 4.6 mm ID with guard column adapter EC (REF 721359) |