Learn More
Macherey-Nagel™ Nucleosil-Nucleodex γ-PM CC Guard Columns Cartridges
Brand:  Macherey-Nagel™ 721466.40
Additional Details : Weight : 0.01000kg
Product Code. 10206561
  • € 124.00 / Each
Estimated Shipment: 20-06-2024
to see stock.
Specifications
1 Pack
8 mm
CC Guard Columns Cartridges
EC columns of 4 and 4.6 mm ID with guard column adapter EC (REF 721359)
5 μm
100Å
4 mm
Nucleosil-Nucleodex γ-PM