Learn More
Macherey-Nagel™ Nucleosil™ SB CC Guard Columns
Product Code. 12728142 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Pack of 3
Product Code. 12728142
Brand: Macherey-Nagel™ 721885.40
Only null left
Specifications
Carbon Load 10%
pH 2 to 8
Particle Size 5 μm
Length (Metric) 8 mm
Pore Size 100Å
Column Capacity ∽ 1 Meq/g
Type HPLC CC Guard Column
Diameter (Metric) Inner 4 mm
Stationary Phase NUCLEOSIL™ SB
Mobile Phase Compatibility Diammonium phosphate
Solvents Diammonium phosphate
Packing Material Silica
Crosslinking Quaternary ammonium
Column Format Guard Column
For Use With (Application) HPLC
Technique HPLC
Particle Shape Spherical
Hydrophobicity Yes
Functionality/Interaction Type Ion Exchange
USP Type L14