Learn More
Macherey-Nagel™ Nucleosil™ SiOH CC Guard Columns
Product Code. 12718142 Shop All Macherey-Nagel Products
Change view
Click to view available options
Diameter (Metric) Inner:
4 mm
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
12718142 4 mm Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12718142 Supplier Macherey-Nagel™ Supplier No. 721872.40
Estimated Shipment: 19-08-2026
Specifications
pH 2 to 8
Particle Size 5 μm
Length (Metric) 8 mm
Pore Size 100Å
Type HPLC CC Guard Column
Diameter (Metric) Inner 4 mm
Stationary Phase NUCLEOSIL™ SiOH
Mobile Phase Compatibility n-heptane
Solvents n-heptane
Packing Material Silica
Column Format Guard Column
For Use With (Application) HPLC
Technique HPLC
Particle Shape Spherical
Hydrophobicity Yes
Functionality/Interaction Type Polar
USP Type L3