missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 8 Blood QuickPure, 8-well kit for quick DNA extraction from blood

Product Code. p-9281005
Click to view available options
No. of Reactions:
480 Preps
96 Preps
Unit Size:
480 reactions
96 reactions
This item is not returnable. View return policy

Product Code. 12738522

Brand: Macherey-Nagel™ 740666

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ultra-fast isolation of highly concentrated DNA from blood in a flexible 8‐well strip format.

Flexible handling of multiples of 8 samples is achieved by use of a reusable column holder. Parallel lysis of large sample numbers can be performed in standard microcentrifuge tubes or in 96 well blocks alternatively. NucleoSpin™ Blood QuickPure binding strips are to be used in a centrifuge with deep buckets allowing processing of NucleoSpin™ 8 well strips stacked on 96 well blocks. The method is based on a specially treated silica membrane which speeds up the procedure by a combined washing/drying step. The number of washing and drying steps is reduced from 3 to only 1, therefore hands-on time is minimised.Lysis is achieved by incubation of whole blood in a solution containing large amounts of chaotropic ions in the presence of proteinase K. Appropriate conditions for binding of DNA to the silica membrane of the NucleoSpin™ Blood QuickPure binding strips are created by addition of ethanol to the lysate. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer and can be used directly for PCR, Southern blotting or any kind of enzymatic reaction. The kit allows isolation of 4 μg to 6 μg of pure genomic DNA from 200 μL whole blood with an A260/280 ratio between 1.60 and 1.90 and a typical concentration of 40 ng/μL to 60 ng/μL.

Kit components: NucleoSpin™ Blood QuickPure binding strips, buffers, proteinase K, accessories.

  • Silica membrane technology
  • Manual processing
  • Yield: 4 to 6 μg
  • Sample material: human or animal whole blood (200 μL/300 μL)
  • Elution volume: 7.5 to 100 μL
  • Binding capacity: 60 μg
  • Preparation time: 60 min./12 strips

Specifications

Content And Storage 18°C to 25°C
Format 8-well Strip
For Use With (Application) Isolation of DNA
No. of Reactions 96 Preps
compliance-icons compliance-icons
Hazard Category
  • Skin sensitiser Category 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.