Learn More
Macherey-Nagel™ NucleoSpin™ 8 PCR Clean-up Core Kit, 8-well kit for PCR clean up
Click to view available options
:
No. of Reactions: 8
Specifications
Specifications
| No. of Reactions | 8 |
| Product Type | NucleoSpin 8 PCR |
| Sample Volume | <100 μL |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- MIXTURE LIST-Contains : guanidinium thiocyanate