missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 8 Plant II, 8-well kit for DNA from plants
Specifications
Specifications
| Content And Storage | 18°C to 25°C |
| Format | 8-well Strip |
| Sample Type | Plants |
| For Use With (Application) | Isolation of DNA |
| No. of Reactions | 480 Preps |
| Sample Volume | 20–100 mg wet weight |
| Size | 50 bp–approx. 50 kbp |
| Target | DNA |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction