missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Trace, 96-well kit for DNA from forensic samples

Product Code. 12728552
Change view
Click to view available options
No. of Reactions:
192 Preps
384 Preps
Unit Size:
192 reactions
384 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12728552 384 Preps 384 reactions
12718552 192 Preps 192 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12728552 Supplier Macherey-Nagel™ Supplier No. 740726.4
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Isolation of DNA from forensic samples in flexible 96‐well strip format.

Lysis is achieved by incubation of samples in a solution containing chaotropic ions in the presence of proteinase K. If subsequent separation of the lysate from the sample material is difficult, the lysate can be separated from the forensic material under vacuum or centrifugation by use of the NucleoSpin™ Trace filter plate (not included in the kits). Appropriate conditions for binding of DNA to the silica membrane in the NucleoSpin™ Trace binding strip/binding plate are created by addition of isopropanol to the lysate. The binding process is reversible and specific to nucleic acids. Contaminants are removed by two washing steps with ethanolic buffer. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer.Typically yields of 1 to 2 μg genomic DNA can be purified from buccal swabs. The final concentration of eluted DNA is 10 to 20 ng/μL (depending on elution buffer volume). Typically, A260/280 is 1.8 to 1.9. The obtained DNA can be used directly as template for PCR.

Kit components

NucleoSpin™ Trace binding strips, MN wash plates, MN square well blocks, rack with tube strips, cap strips, self-adhering foil, buffers, proteinase K. NucleoSpin™ 96 Trace - NucleoSpin™ Trace binding plates, MN wash plates, MN square well blocks, racks with tube strips, cap strips, buffers, proteinase K.

  • Suitable for manual and automated processing, processing possible under vacuum or by centrifugation
  • 96 well plate format
  • Typical yield: 1 to 2 μg DNA from buccal swabs
  • Elution volume: 50 to 100 μL
  • Silica membrane technology
  • No use of organic solvents
  • Highly pure nucleic acids suitable for all common downstream applications
  • Preparation time: 70 min./plate
  • Binding capacity: 20 μg

Specifications

Content And Storage 18°C to 25°C
Format 96-well Plate
For Use With (Application) Isolation of DNA
No. of Reactions 384 Preps
compliance-icons compliance-icons
Produktbeteckning
  • NucleoSpin 96 Trace (4x96)
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Kompletterande information
  • MIXTURE LIST-Contain: proteinase K
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.