missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 Virus, 96-well kit for viral RNA/DNA purification

Product Code. p-9281007
Click to view available options
No. of Reactions:
192 Preps
384 Preps
Unit Size:
1 set
384 reactions
This item is not returnable. View return policy

Product Code. 12796533

Brand: Macherey-Nagel™ 740691.2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Time saving parallel isolation of viral RNA / DNA from biological fluids in proven 96‐well plate format.

Designed for the simultaneous purification of viral RNA and DNA. The kit combines the selectivity of well established silica membrane binding of nucleic acids with high throughput 96-well format. With the NucleoSpin™ 96 Virus method, RNA viruses are quickly and efficiently lysed by Lysis Buffer RAV1 which is a highly concentrated GITC solution. Compared to RNA viruses, DNA viruses (e.g., HBV) are usually more difficult to isolate and require a digestion of samples with Proteinase K which is provided in the kit. Lysis buffer and ethanol create appropriate conditions for binding of nucleic acids to the silica membrane of the NucleoSpin™ Virus Binding Plate. Carrier RNA included in Lysis Buffer RAV1 improves binding and recovery of low concentrated viral RNA/DNA. Contaminations (potential PCR inhibitors) like salts, metabolites, and soluble macromolecular cellular components are removed in washing steps with ethanolic Wash Buffer RAW and Buffer RAV3. The purified viral nucleic acids can be eluted in low salt buffer or water and are ready-to-use in subsequent downstream applications like RT-PCR or PCR.

  • Silica membrane technology
  • Sample material: Cell-free biological fluid, Plasma, Serum
  • Fragment size: 100 bp to 50 kbp
  • Theoretical binding capacity: 40 μL
  • Elution volume: 70 to 100 μL
  • Preparation time: 60 min./plate

Specifications

Content And Storage 18°C to 25°C
Format 96-well Plate
For Use With (Application) Isolation of viral nucleic acids
No. of Reactions 192 Preps
Sample Volume <150 μL
Target Viral RNA and DNA
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.