Learn More
Macherey-Nagel™ NucleoSpin™ DNA Forensic, Mini kit for DNA from forensic traces
Product Code. 15777307
Change view
Click to view available options
No. of Reactions:
10 Preps
250 Preps
50 Preps
Unit Size:
10 reactions
250 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
15777307 10 Preps 10 reactions
15797307 50 Preps 50 reactions
15787307 250 Preps 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15777307 Supplier Macherey-Nagel™ Supplier No. 740840.10
Spin technology based isolation of genomic DNA from forensic samples (e.g., casework samples, contact traces). Complete kits (spin columns, buffers, collection tubes, liquid proteinase K, reducing agent TCEP) available in several quantities.
NucleoSpin™ DNA Forensic Kits
  • Excellent genomic DNA purity from all casework samples
  • Procedure is based on reversible adsorption of nucleic acids to the silica membrane
  • Sample sources include buccal swabs or small tissue/cell samples (dried blood spots, trace and touch-samples or cigarette filters)
  • Uniform buffer system for single sample analysis and for high throughput analysis
  • Ethylene oxide-treated plastic materials
  • Conformity to ISO 18385 for doubtless DNA profiling
  • Applications: STR analysis, qPCR, any kind of enzymatic reaction
Specifications
Concentration 10 to 30 ng/μL
Content And Storage Room temperature (18-25 °C)
Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Sample Type Casework Samples, Contact Traces (e.g., Dried Blood Spots, Cigarette Filters, Swabs)
Efficiency Binding capacity: 7 μg
Elution Volume Elution volume: 50-100 μL
Final Product Type Genomic DNA
For Use With (Equipment) Vortex mixer
For Use With (Application) Isolation of DNA from forensic samples
Includes NucleoSpin DNA Forensic columns, buffers, collection tubes (2 mL), liquid proteinase K, reducing agent TCEP
No. of Reactions 10 Preps
Yield 1-3 μg (from buccal swabs)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • NucleoSpin DNA Forensic (10)
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H413-May cause long lasting harmful effects to aquatic life.
Supplemental information
  • MIXTURE LIST-Contain: guanidium thiocyanate, 2-propanol, guanidium hydrochloride,ethanol, proteinasek