missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ DNA Lipid Tissue, Mini kit for DNA from lipid rich tissue
Click to view available options
:
50 Preps
Description
Isolation of genomic DNA from lipid-rch tissue
- Fast and convenient procedure combining enzymatic lysis and bead disruption
- Special buffer for efficient removal of lipids, which influence the chemistry of most buffers
- Suitable for a wide variety of lipid-rich samples (e.g., brain, adipose tissue, fatty fish tissue)
- High quality DNA from fresh or frozen samples
- Bead Tubes included for efficient lysis of lipids, which can interfere with tissue disruption
- Compatible with common disruption devices (e.g. Vortex-Genie 2 with MN Bead Tube Holder)
- Typical downstream applications: multiplex PCR, Southern blotting, enzymatic reactions
Specifications
Specifications
| Content And Storage | Room temperature (18 - 25 °C) |
| Format | Mini Spin Column |
| Isolation Technology | Silica MembraneTechnology and Mechanical (Bead Tube) Disruption |
| Sample Type | Lipid Rich Tissue |
| Efficiency | Binding capacity: 60 μg |
| Elution Volume | Elution volume: 25 - 200 μL |
| For Use With (Equipment) | Disruption device (e.g. Vortex-Genie 2 with MN Bead Tube Holder), Centrifuge, Vortex mixer |
| For Use With (Application) | Isolation of genomic DNA from lipid-rich tissue |
| Includes | NucleoSpin™ DNA Lipid Tissue Columns, NucleoSpin™ Bead Tubes Type D, Collection Tubes (2 mL), Buffers |
| No. of Reactions | 50 |
| Show More |
Safety and Handling
Product Identifier
- NucleoSpin DNA Lipid Tissue (50)
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- MIXTURE LIST-Contains : 2-propanol, guanidine hydrochloride, gyanidium thiocyanate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction