missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nucleotide binding protein like Antibody (OTI5D5), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73094AF532
This item is not returnable.
View return policy
Description
Nucleotide binding protein like Monoclonal specifically detects Nucleotide binding protein like in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Nucleotide binding protein like | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NUBPL | |
| Recombinant protein fragment corresponding to amino acids 1-250 of human Nucleotide binding protein like (NP_079428) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5D5 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| C14orf127, FLJ12660, huInd1IND1 homolog, IND1chromosome 14 open reading frame 127, iron-sulfur protein NUBPL, iron-sulfur protein required for NADH dehydrogenase, nucleotide binding protein-like, Nucleotide-binding protein-like | |
| Mouse | |
| 0.1 mL | |
| 80224 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction