Learn More
NUDC (phospho S326) Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human NUDC.
Brand: Abnova PAB29584.100uL
Description
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein (see DYNC1H1; MIM 600112) (Aumais et al., 2003 [PubMed 12679384]).[supplied by OMIM]
Specifications
| NUDC (phospho S326) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of human NUDC. | |
| In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| NUDC | |
| A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S326 of human NUDC. | |
| 100 μL | |
| Primary | |
| NUDC (phospho S326) polyclonal antibody detects endogenous levels of human NUDC only when phosphorylated at serine 326. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
| HNUDC/MNUDC/NPD011 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10726 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.