missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10681-1-AP-150UL
This item is not returnable.
View return policy
Description
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein.Specifications
| NUDC | |
| Polyclonal | |
| Unconjugated | |
| NUDC | |
| HNUDC, MNUDC, Nuclear migration protein nudC, NUDC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10726 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y266 | |
| NUDC | |
| NUDC Fusion Protein Ag0998 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction