Learn More
NUDC Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10681-1-AP-20UL
 View more versions of this product
Product Code. 16817223
  • € 145.00 / 20µL
Description
NudC was first identified as a regulator of nuclear movement in the asexual reproductive cycle of the filamentous fungus Aspergillus nidulans. Human NUDC is a nuclear movement protein that associates with dynein.
Specifications
Specifications
NUDC
Polyclonal
Unconjugated
NUDC
HNUDC, MNUDC, Nuclear migration protein nudC, NUDC
Rabbit
Antigen Affinity Chromatography
RUO
10726
-20°C
Liquid
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y266
NUDC
NUDC Fusion Protein Ag0998