missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NUDCD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546769
This item is not returnable.
View return policy
Description
Peptide sequence: YAADPWVQDQ MQRKLTLERF QKENPGFDFS GAEISGNYTK GGPDFSNLEK.
NUDCD2 may regulate the LIS1/dynein pathway by stabilizing LIS1 with Hsp90 chaperone.
Specifications
| NUDCD2 | |
| Polyclonal | |
| Unconjugated | |
| Nudcd2 | |
| D11Ertd603e; NudC domain containing 2; nudC domain-containing protein 2; Nudcd2; NudCL2; NudC-like protein 2; RGD1307203 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 134492 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WVJ2 | |
| Nudcd2 | |
| Synthetic peptide directed towards the C-terminal of human NUDCD2 (aa 108-157). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction