missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nudel Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97160-100ul
This item is not returnable.
View return policy
Description
Nudel Polyclonal antibody specifically detects Nudel in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Nudel | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate | |
| DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel | |
| Produced in rabbits immunized with E. coli-derived Human Nudel fragment. | |
| 100 μg | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction | |
| 81565 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction