missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nudel Antibody (OTI5E11), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73104APC
This item is not returnable.
View return policy
Description
Nudel Monoclonal specifically detects Nudel in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Nudel | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| NDEL1 | |
| Full length recombinant protein of human Nudel (NP_001020750) produced in HEk293 cell. | |
| 0.1 mL | |
| 81565.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI5E11 | |
| Flow Cytometry | |
| DKFZp451M0318, EOPA, MITAP1mitosin-associated protein MITAP1, Mitosin-associated protein 1, nuclear distribution protein nudE-like 1, nudE nuclear distribution gene E homolog (A. nidulans)-like 1, nudE nuclear distribution gene E homolog like 1, nudE nuclear distribution gene E homolog like 1 (A. nidulans), NUDELendooligopeptidase A, NUDE-like protein, Protein Nudel | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction