missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nudel Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596422

Product Code. 16539261

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MDGEDIPDFS SLKEETAYWK ELSLKYKQSF QEARDELVEF QEGSRELEAE LEAQLVQAEQ RNRDLQADNQ RLKYEVEALK EKLEHQYAQS YKQVSVLEDD LSQTRAIKEQ LHKYVRELEQ ANDDLERAKR ATIVSLEDFE QRLNQAIERN AFLESELDEK ESLLVSVQRL KDEARDLRQE LAVRERQQEV TRKSAPSSPT LDCEKMDSAV QASLSLPATP VGKGTENTFP SPKAIPNGFG TSPLTPSARI SALNIVGDLL RKVGALESKL AACRNFAKDQ ASRKSYISGN VNCGVLNGNG TKFSRSGHTS FFDKGAVNGF DPAPPPPGLG SSRPSSAPGM LPLSV; Positive Samples: U-87MG, Mouse brain; Cellular Location: Chromosome, Cytoplasm, centromere, centrosome, cytoskeleton, kinetochore, microtubule organizing center, spindle.

This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nudel
Polyclonal
Unconjugated
Ndel1
2600006O07Rik; ATP-sensitive inward rectifier potassium channel 15; DKFZp451M0318; endooligopeptidase A; EOPA; Inward rectifier K(+) channel Kir4.2; Kcnj15; Kir4.2; LIS1-interacting protein NUDEL; LIS1-interacting protein NUDEL; endooligopeptidase A; MITAP1; Mitosin-associated protein 1; mitosin-associated protein MITAP1; mNudel; mNudE-L; NDE1L1; NDE2; Ndel1; nuclear disribution gene E like; nuclear distribution gene E-like homolog 1; nuclear distribution gene E-like homolog 1 (A. nidulans); nuclear distribution protein nudE-like 1; nudE neurodevelopment protein 1 like 1; nudE neurodevelopment protein 1-like 1; nudE nuclear distribution E homolog-like 1; nudE nuclear distribution gene E homolog like 1; nudE nuclear distribution gene E homolog-like 1; Nude2; NUDEL; NUDE-like protein; Potassium channel, inwardly rectifying subfamily J member 15; potassium channel, inwardly rectifying subfamily J, member 15; potassium inwardly-rectifying channel, subfamily J, member 15; Protein mNudE-like; Protein Nudel; unnamed protein product
Rabbit
Affinity Chromatography
RUO
81565, 83431
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.73 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9ERR1, Q9GZM8
Ndel1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-345 of human NUDEL/NUDEL/NDEL1 (NP_1104351).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.