missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46738
This item is not returnable.
View return policy
Description
NUDT10 Polyclonal antibody specifically detects NUDT10 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| NUDT10 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| Q96G61 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| APS2DIPP3A, Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 3-alpha, diphosphoinositol polyphosphate phosphohydrolase type 3 alpha, DIPP3a, DIPP-3-alpha, DIPP3-alpha, EC 3.6.1.-, EC 3.6.1.52, hDIPP3alphahAps2, Nucleoside diphosphate-linked moiety X motif 10, nudix (nucleoside diphosphate linked moiety X)-type motif 10, Nudix motif 10, P6-hexaphosphate hydrolase 3-alpha | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EVLLVSSSRYPDRWIVPGGGMEPEE | |
| 0.1 mL | |
| Signal Transduction | |
| 170685 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction