missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT10 Antibody (OTI1A6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46268
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
NUDT10 Monoclonal specifically detects NUDT10 in Human samples. It is validated for Western Blot.
Specifications
| NUDT10 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| APS2DIPP3A, Diadenosine 5'-5'''-P1, diphosphoinositol polyphosphate phosphohydrolase 3-alpha, diphosphoinositol polyphosphate phosphohydrolase type 3 alpha, DIPP3a, DIPP-3-alpha, DIPP3-alpha, EC 3.6.1.-, EC 3.6.1.52, hDIPP3alphahAps2, Nucleoside diphosphate-linked moiety X motif 10, nudix (nucleoside diphosphate linked moiety X)-type motif 10, Nudix motif 10, P6-hexaphosphate hydrolase 3-alpha | |
| Mouse | |
| 18.3 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 170685 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI1A6 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| NUDT10 | |
| Full length human recombinant protein of human NUDT10 (NP_694853) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction