Learn More
NUDT10 Monoclonal Antibody (OTI2D9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803923
Encompass_Preferred
 View more versions of this product
Product Code. 17573223
  • € 540.87 / 100µL
Description
Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphates, with Ap6A and Ap5A being the preferred substrates. The major reaction products are ADP and p4a from Ap6A and ADP and ATP from Ap5A. Also able to hydrolyze 5-phosphoribose 1-diphosphate.
Specifications
Specifications
NUDT10
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8NFP7
NUDT10
Full length human recombit protein of human NUDT10 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2D9
Unconjugated
NUDT10
APS2, DIPP3-alpha, DIPP3a
Mouse
Affinity Chromatography
RUO
170685
-20° C, Avoid Freeze/Thaw Cycles