Learn More
NUDT10 Mouse anti-Human, Clone: OTI2G8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804113
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cleaves a beta-phosphate from the diphosphate groups in PP-InsP5 (diphosphoinositol pentakisphosphate), suggesting that it may play a role in signal transduction. Also able to catalyze the hydrolysis of dinucleoside oligophosphates, with Ap6A and Ap5A being the preferred substrates. The major reaction products are ADP and p4a from Ap6A and ADP and ATP from Ap5A. Also able to hydrolyze 5-phosphoribose 1-diphosphate.Specifications
| NUDT10 | |
| Monoclonal | |
| Unconjugated | |
| NUDT10 | |
| APS2, DIPP3-alpha, DIPP3a | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 170685 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2G8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q8NFP7 | |
| NUDT10 | |
| Full length human recombit protein of human NUDT10 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.