missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT12 Monoclonal Antibody (OTI5B6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805570
This item is not returnable.
View return policy
Description
Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Nucleotides are involved in numerous biochemical reactions and pathways within the cell as substrates, cofactors, and effectors. Nudix hydrolases, such as NUDT12, regulate the concentrations of individual nucleotides and of nucleotide ratios in response to changing circumstances.Specifications
| NUDT12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQG2 | |
| NUDT12 | |
| Human recombit protein fragment corresponding to amino acids 1-251 of human NUDT12 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5B6 | |
| Unconjugated | |
| NUDT12 | |
| nudix hydrolase 12 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 83594 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction