missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT13 Antibody (1A9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00025961-M02
This item is not returnable.
View return policy
Description
NUDT13 Monoclonal antibody specifically detects NUDT13 in Human samples. It is validated for ELISA, ELISA
Specifications
| NUDT13 | |
| Monoclonal | |
| Unconjugated | |
| NP_056985 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 25961 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| ELISA, Sandwich ELISA | |
| 1A9 | |
| In 1x PBS, pH 7.4 | |
| DKFZp586P2219, EC 3.-, nucleoside diphosphate-linked moiety X motif 13, nudix (nucleoside diphosphate linked moiety X)-type motif 13, Nudix motif 13, Protein KiSS-16 | |
| NUDT13 (NP_056985.3, 2 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SLYCGIACRRKFFWCYRLLSTYVTKTRYLFELKEDDDACKKAQQTGAFYLFHSLAPLLQTSAHQYLAPRHSLLELERLLGKFGQDAQRIEDSVLIGCSEQQEAWFALDL | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction