missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT18 Antibody (OTI5D7), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73110MFV500
This item is not returnable.
View return policy
Description
NUDT18 Monoclonal antibody specifically detects NUDT18 in Human samples. It is validated for Flow Cytometry
Specifications
| NUDT18 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| OTI5D7 | |
| Flow Cytometry | |
| EC 3.6.1, EC 3.6.1.-, FLJ22494, nucleoside diphosphate-linked moiety X motif 18, nudix (nucleoside diphosphate linked moiety X)-type motif 18, Nudix motif 18 | |
| Full length human recombinant protein of human NUDT18(NP_079091) produced in HEK293T cell. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 79873 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction