missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT18 Antibody (OTI5D7), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73110PE
This item is not returnable.
View return policy
Description
NUDT18 Monoclonal specifically detects NUDT18 in Human samples. It is validated for Flow Cytometry.
Specifications
| NUDT18 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| NUDT18 | |
| Full length recombinant protein of human NUDT18 (NP_079091) produced in HEK293T cells. | |
| 0.1 mL | |
| 79873.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI5D7 | |
| Flow Cytometry | |
| EC 3.6.1, EC 3.6.1.-, FLJ22494, nucleoside diphosphate-linked moiety X motif 18, nudix (nucleoside diphosphate linked moiety X)-type motif 18, Nudix motif 18 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction