missing translation for 'onlineSavingsMsg'
Learn More

NUDT18 Monoclonal Antibody (OTI5D7), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA503847S

Encompass_Preferred

 View more versions of this product

Product Code. 17500162

  • € 242.00 / 30µL
Only null left
Estimated Shipment: 26-01-2026
Add to basket
This item is not returnable. View return policy

Description

Description

NUDT18 Monoclonal antibody specifically detects NUDT18 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

NUDT18
Monoclonal
0.55 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q5SW96
Mouse
Affinity chromatography
RUO
26119
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI5D7
Unconjugated
LDLRAP1
2-hydroxy-dADP phosphatase; 7,8-dihydro-8-oxoguanine phosphatase; 8-oxo-dGDP phosphatase NUDT18; MTH3; MutT homolog 3; mutT human homolog 3; nucleoside diphosphate-linked moiety X motif 18; nudix (nucleoside diphosphate linked moiety X)-type motif 18; nudix hydrolase 18; nudix motif 18; NUDT18
Full length human recombinant protein of human LDLRAP1 produced in HEK293T cell.
30 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.