missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT18 Monoclonal Antibody (OTI5F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503865
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the Nudix hydrolase family. Nudix hydrolases eliminate potentially toxic nucleotide metabolites from the cell and regulate the concentrations and availability of many different nucleotide substrates, cofactors, and signaling molecules. This protein contains a Nudix hydrolase domain and hydrolyzes oxidized forms of guanosine and deoxyguanosine diphosphates.Specifications
| NUDT18 | |
| Monoclonal | |
| 0.79 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6ZVK8 | |
| NUDT18 | |
| Full length human recombit protein of human NUDT18 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5F9 | |
| Unconjugated | |
| NUDT18 | |
| MTH3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 79873 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction