missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT21 Monoclonal Antibody (OTI13H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806779
This item is not returnable.
View return policy
Description
NUDT21 is one subunit of a cleavage factor required for 3′ RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3′ end processing complex and facilitates the recruitment of other processing factors.The protein encoded by this gene is one subunit of a cleavage factor required for 3′ RNA cleavage and polyadenylation processing. The interaction of the protein with the RNA is one of the earliest steps in the assembly of the 3′ end processing complex and facilitates the recruitment of other processing factors. This gene encodes the 25kD subunit of the protein complex, which is composed of four polypeptides.Specifications
| NUDT21 | |
| Monoclonal | |
| Unconjugated | |
| NUDT21 | |
| CFIM25, CPSF5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11051 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI13H1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43809 | |
| NUDT21 | |
| Human recombit protein fragment corresponding to amino acids 2-227 of human NUDT21 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction