missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD13 (WM15), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65186-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD13 antibody from Proteintech is a mouse monoclonal antibody generated with human aml cells of human CD13. This antibody recognizes human, non-human primates antigen. The CD13 antibody has been validated for the following applications: IF/ICC, FC analysis.Specifications
| CD13 | |
| Monoclonal | |
| 0.5 mg/mL | |
| APN; CD13; LAP1; PEPN; gp150; p150 | |
| Human AML cells | |
| 500 μg | |
| Primary | |
| Human, Non-human Primate | |
| Flow Antibody | |
| IgG1 κ |
| Immunocytochemistry/Immunofluorescence, Flow Cytometry | |
| WM15 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 290 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction