missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT22 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | NUDT22 |
|---|---|
| Clone | 3C3H9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235605
|
Proteintech
67921-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233825
|
Proteintech
67921-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NUDT22 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NUDT22. This antibody recognizes human, rat antigen. The NUDT22 antibody has been validated for the following applications: WB, ELISA, FC (Intra) analysis.Specifications
| NUDT22 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 84304 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 3C3H9 | |
| Western Blot, ELISA, Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| MGC13045 | |
| IgG2b | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title