missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT6 Antibody (OTI9D12), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73121PE
This item is not returnable.
View return policy
Description
NUDT6 Monoclonal specifically detects NUDT6 in Human samples. It is validated for Flow Cytometry.
Specifications
| NUDT6 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| NUDT6 | |
| Full length recombinant protein of human NUDT6 (NP_009014) produced in HEK293T cells. | |
| 0.1 mL | |
| 11162.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI9D12 | |
| Flow Cytometry | |
| Antisense basic fibroblast growth factor, ASFGF2, bFGF, FGF2ASFGF-2, FGF-AS, gfg, gfg-1, nucleoside diphosphate-linked moiety X motif 6, nudix (nucleoside diphosphate linked moiety X)-type motif 6, Nudix motif 6, nudix-type motif 6, Protein GFG | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction