missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT6 Monoclonal Antibody (OTI10A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502342S
This item is not returnable.
View return policy
Description
NUDT6 Monoclonal antibody specifically detects NUDT6 in Canine, Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| NUDT6 | |
| Monoclonal | |
| 0.46 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| B6V8E6, P35222, Q9WU82 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1499, 477032, 84353 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI10A9 | |
| Unconjugated | |
| CTNNB1 | |
| antisense basic fibroblast growth factor; Antisense basic fibroblast growth factor B; anti-sense basic fibroblast growth factor B; ASFGF2; Asfgf2b; FGF2AS; FGF-AS; Gfg; GFG1; GFG-1; Nucleoside diphosphate-linked moiety X motif 6; nudix (nucleoside diphosphate linked moiety X)-type motif 6; nudix hydrolase 6; nudix motif 6; Nudt6; Protein GFG | |
| Full length human recombinant protein of human CTNNB1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction