missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT6 Monoclonal Antibody (OTI10A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502342
This item is not returnable.
View return policy
Description
NUDT6 (nudix (nucleoside diphosphate linked moiety X)-type motif 6), also known as Antisense basic fibroblast growth factor or GFG-1, is a member of the Nudix hydrolase family of pyrophosphatases. Nudix hydrolases contain a characteristic Nudix domain and are responsible for catalyzing the hydrolysis of nucleoside diphosphate derivatives. The gene encoding NUDT6 is a FGF-2 gene antisense transcript, and NUDT6 is believed to regulate FGF-2 expression. FGF-2 is a mutifunctional heparin-binding growth factor important to angiogenesis, neuroectoderm development and wound healing. NUDT6 is expressed as two isoforms produced by alternative splicing.Specifications
| NUDT6 | |
| Monoclonal | |
| 0.46 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P53370 | |
| NUDT6 | |
| Full length human recombit protein of human NUDT6 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI10A9 | |
| Unconjugated | |
| NUDT6 | |
| ASFGF2, FGF-AS, FGF2AS, GFG-1, GFG1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 11162 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction