missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT9 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97230V
This item is not returnable.
View return policy
Description
NUDT9 Polyclonal antibody specifically detects NUDT9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NUDT9 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| ADP-ribose diphosphatase, ADP-ribose phosphohydrolase, ADP-ribose pyrosphosphatase NUDT9, ADPR-PPase, EC 3.6.1.13, MGC3037, mitochondrial, nucleoside diphosphate linked moiety X-type motif 9, nudix (nucleoside diphosphate linked moiety X)-type motif 9, Nudix motif 9, NUDT10 | |
| Produced in rabbits immunized with E. coli-derived Human NUDT9 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 53343 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction