missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT9 Antibody (OTI 7F12), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73124H
This item is not returnable.
View return policy
Description
NUDT9 Monoclonal specifically detects NUDT9 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NUDT9 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| NUDT9 | |
| Full length recombinant protein of human NUDT9 (NP_076952) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot | |
| OTI 7F12 | |
| Western Blot | |
| ADP-ribose diphosphatase, ADP-ribose phosphohydrolase, ADP-ribose pyrosphosphatase NUDT9, ADPR-PPase, EC 3.6.1.13, MGC3037, mitochondrial, nucleoside diphosphate linked moiety X-type motif 9, nudix (nucleoside diphosphate linked moiety X)-type motif 9, Nudix motif 9, NUDT10 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 53343.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction