missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuf2 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32211FR
This item is not returnable.
View return policy
Description
Nuf2 Polyclonal antibody specifically detects Nuf2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Nuf2 | |
| Polyclonal | |
| Western Blot | |
| cancer/testis antigen 106, CDCA1hsNuf2, cell division cycle associated 1, Cell division cycle-associated protein 1, CT106, hNuf2, kinetochore protein Nuf2, NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae), NUF2RhNuf2R | |
| The immunogen this antibody was made to, maps to a region between residue 414 to 464 of human Nuclear Fallout 2 using the numbering given in entry NP_663735.2 (GeneID 83540). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 83540 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction