Learn More
Invitrogen™ NUFIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110050
Description
Immunogen sequence: DMSLTSAAVE QIKTSLFIYP SNMQTMLLST AQVDLPSQTD QQNLGDIFQN QWGLSFINEP SAGPETVTGK SSEHKVMEVT FQGEYPATLV SQGAEIIPSG TEHPVFPKAY ELEKRTSPQV LGSILKSGTT SESGALSLEP SHIGDLQKAD TSSQGALVFL SKDYEIESQN PLASPTNTLL GSAKEQRYQR GLERNDSWGS FDLRAAIVYH TKEMESIWNL QKQDPKRIIT YNEAMDSPDQ.
NUFIP2 binds RNA.
Specifications
| NUFIP2 | |
| Polyclonal | |
| Unconjugated | |
| Nufip2 | |
| 1110001M19Rik; 182-FIP; 82 kDa FMRP-interacting protein; 82-FIP; 82-kD FMRP Interacting Protein; 9530056D24Rik; AI256692; AI461731; Cell proliferation-inducing gene 1 protein; FIP-82; FMRP-interacting protein 2; Fragile X Mental Retardation Protein; im:7136583; KIAA1321; LOW QUALITY PROTEIN: nuclear fragile X mental retardation-interacting protein 2; mKIAA1321; nuclear FMR1 interacting protein 2; nuclear FMR1 interacting protein 2; nuclear fragile X mental retardation-interacting protein 2; nuclear fragile X mental retardation protein interacting protein 2; nuclear fragile X mental retardation-interacting protein 2; NUFIP2; NUFIP2, FMR1 interacting protein 2; PIG1; proliferation-inducing gene 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57532, 681371, 68564 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.76 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5F2E7, Q7Z417 | |
| Nufip2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 456-695 of human NUFIP2 (NP_065823.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.