missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NuMA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 18 publications
€ 224.00 - € 531.00
Specifications
| Antigen | NuMA |
|---|---|
| Dilution | Western Blot 1:1000, Simple Western 1:500, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18076953
|
Novus Biologicals
NB500-174SS |
0.025 mL |
€ 224.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18728953
|
Novus Biologicals
NB500-174 |
0.1 mL |
€ 531.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NuMA Polyclonal specifically detects NuMA in Human, Mouse, Rat, Mammal, Marsupial, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| NuMA | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Mitotic Regulators | |
| Whole antisera with 0.02% Sodium Azide | |
| centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic apparatus protein 1, NUMA, NuMA protein, SP-H antigen, structural nuclear protein | |
| NUMA1 | |
| IgG | |
| Unpurified | |
| Reactive to most mammalian species tested. |
| Western Blot 1:1000, Simple Western 1:500, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Mammalia, Marsupial, Primate | |
| Q14980 | |
| 4926 | |
| Full-length recombinant human NuMA expressed in insect Sf9 cells using baculovirus expression system, then purified to near homogeneity prior to immunization. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title