Learn More
NuMA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-74636
Product Code. 18797153
  • € 603.00 / 0.10mL
Description
NuMA Polyclonal specifically detects NuMA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Immunohistochemistry, Immunocytochemistry
centrophilin stabilizes mitotic spindle in mitotic cells, nuclear mitotic apparatus protein 1, NUMA, NuMA protein, SP-H antigen, structural nuclear protein
238.26 kDa
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:500 - 1:2000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000
NUMA1