missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Numb Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-178MFV500
This item is not returnable.
View return policy
Description
Numb Polyclonal antibody specifically detects Numb in Human, Mouse, Chicken samples. It is validated for Western Blot, Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Numb | |
| Polyclonal | |
| Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence | |
| h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
| A synthetic peptide made to a C-terminal region of mouse NUMB (between residues 600-653). [UniProt# Q9QZS3] | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 8650 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction