missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Numb Antibody (OTI4F3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73129AF647
This item is not returnable.
View return policy
Description
Numb Monoclonal specifically detects Numb in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Numb | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NUMB | |
| Full length recombinant protein of human Numb (NP_003735) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| OTI4F3 | |
| Western Blot, Immunohistochemistry | |
| h-Numb, numb (Drosophila) homolog, numb homolog (Drosophila), protein numb homolog, Protein S171, S171 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 8650.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction