missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUMB Monoclonal Antibody (OTI1E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802140
This item is not returnable.
View return policy
Description
Protein NUMB homolog is an evolutionarily conserved protein that controls the differentiation of neuronal progenitor cells. Recent studies have revealed that NUMB functions in clathrin-dependent endocytosis by binding to the AP-2 complex. NUMB is also expressed in postmitotic neurons and plays a role in axonal growth. Neural cells expressing Numb isoforms with short phosphotyrosine-binding (SPTB) domain undergo extensive neurite outgrowth, an effect that can be blocked by voltage-gated Ca2+ channel (VGCC) inhibitor or by Ca2+ chelator.Specifications
| NUMB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49757 | |
| NUMB | |
| Human recombit protein fragment corresponding to amino acids 1-365 of human NUMB produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI1E3 | |
| Unconjugated | |
| NUMB | |
| C14orf41, S171, c14_5527 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8650 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction