missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUMB Monoclonal Antibody (OTI3H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802162S
This item is not returnable.
View return policy
Description
NUMB Monoclonal antibody specifically detects NUMB in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NUMB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P49757 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8650 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H8 | |
| Unconjugated | |
| Numb | |
| c14_5527; C14orf41; h-Numb; mnb; m-Nb; m-numb; Nb; NUMB; NUMB endocytic adaptor protein; numb gene homolog; numb gene homolog (Drosophila); numb homolog; numb homolog (Drosophila); NUMB isoform 3; Numb variant 1; Numb variant 2; NUMB, endocytic adaptor protein; protein numb homolog; Protein S171; PTB domain protein; S171 | |
| Human recombinant protein fragment corresponding to amino acids 1-365 of human NUMB produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction