Learn More
NUMB Mouse anti-Human, Clone: OTI1B5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF802138
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Protein NUMB homolog is an evolutionarily conserved protein that controls the differentiation of neuronal progenitor cells. Recent studies have revealed that NUMB functions in clathrin-dependent endocytosis by binding to the AP-2 complex. NUMB is also expressed in postmitotic neurons and plays a role in axonal growth. Neural cells expressing Numb isoforms with short phosphotyrosine-binding (SPTB) domain undergo extensive neurite outgrowth, an effect that can be blocked by voltage-gated Ca2+ channel (VGCC) inhibitor or by Ca2+ chelator.Specifications
| NUMB | |
| Monoclonal | |
| Unconjugated | |
| NUMB | |
| C14orf41, S171, c14_5527 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8650 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P49757 | |
| NUMB | |
| Human recombit protein fragment corresponding to amino acids 1-365 of human NUMB produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.