missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUMB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18701-1-AP-20UL
This item is not returnable.
View return policy
Description
Four transcript variants encoding different isoforms have been found for this gene. The antibody can bind to all these four isoforms P65, P66, P71, and P72, but can not recognize NUMBL.
Protein NUMB homolog is an evolutionarily conserved protein that controls the differentiation of neuronal progenitor cells. Recent studies have revealed that NUMB functions in clathrin-dependent endocytosis by binding to the AP-2 complex. NUMB is also expressed in postmitotic neurons and plays a role in axonal growth. Neural cells expressing Numb isoforms with short phosphotyrosine-binding (SPTB) domain undergo extensive neurite outgrowth, an effect that can be blocked by voltage-gated Ca2+ channel (VGCC) inhibitor or by Ca2+ chelator.Specifications
| NUMB | |
| Polyclonal | |
| Unconjugated | |
| NUMB | |
| h Numb, NUMB, numb homolog (Drosophila), Protein numb homolog, Protein S171, S171 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18222, 8650 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49757, Q9QZS3 | |
| Numb | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction