missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUMBL Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-88136B
This item is not returnable.
View return policy
Description
NUMBL Polyclonal antibody specifically detects NUMBL in Human samples. It is validated for Western Blot
Specifications
| NUMBL | |
| Polyclonal | |
| Western Blot | |
| CAG3A, CTG3a, numb (Drosophila) homolog-like, numb homolog (Drosophila)-like, numb homolog-like, NUMBLIKE, numb-like protein, NUMB-R, Numb-related protein, NUMBRNBL, TNRC23 | |
| The immunogen recognized by this antibody maps to a region between residue 200 and 250 of human numb (Drosophila) homolog-like using the numbering given in entry NP_004747.1 (GeneID 9253). | |
| 0.1 mL | |
| Cell Biology, Neuroscience, Stem Cells | |
| 9253 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction